CAS RN 3160-59-6
CAS RN 3160-59-6
IUPAC Name:
(2S,3S)-2-{[(benzyloxy)carbonyl]amino}-3-methylpentanoate
Synonyms:
(2S,3S)-2-{[(benzyloxy)carbonyl]amino}-3-methylpentanoate
Molecular Weight (g/mol):
264.3
Molecular Formula:
C14H18NO4
InChi Key:
JSHXJPFZKBRLFU-JQWIXIFHSA-M
SMILES:
CC[C@H](C)[C@H](NC(=O)OCC1=CC=CC=C1)C([O-])=O
1
–
3
of
3
results
1
N-Cbz-L-isoleucine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N-Cbz-L-isoleucine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
N-Cbz-L-isoleucine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.