accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 3160-47-2

H3COONHOOOHO

CAS RN 3160-47-2

IUPAC Name: 2-{[(benzyloxy)carbonyl]amino}-4-methoxy-4-oxobutanoic acid
Synonyms: 2-{[(benzyloxy)carbonyl]amino}-4-methoxy-4-oxobutanoic acid
Molecular Weight (g/mol): 281.26
Molecular Formula: C13H15NO6
InChi Key: PHMBNDDHIBIDRQ-UHFFFAOYNA-N
SMILES: COC(=O)CC(NC(=O)OCC1=CC=CC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)