accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

ClOOHN

CAS RN 3158-91-6

IUPAC Name: 6-chloro-2-oxa-9-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-10-one
Synonyms: 6-chloro-2-oxa-9-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-10-one
Molecular Weight (g/mol): 245.66
Molecular Formula: C13H8ClNO2
InChi Key: CQTGCCLTIDNYHT-UHFFFAOYSA-N
SMILES: ClC1=CC=C2OC3=CC=CC=C3C(=O)NC2=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results