accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 31571-14-9

H3CONOHCH3H3C(E/Z)

CAS RN 31571-14-9

IUPAC Name: 3-(hydroxyimino)-1,7,7-trimethylbicyclo[2.2.1]heptan-2-one
Synonyms: 3-(hydroxyimino)-1,7,7-trimethylbicyclo[2.2.1]heptan-2-one
Molecular Weight (g/mol): 181.24
Molecular Formula: C10H15NO2
InChi Key: YRNPDSREMSMKIY-UHFFFAOYNA-N
SMILES: CC1(C)C2CCC1(C)C(=O)C2=NO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results