accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 315703-52-7

H3CONHNSNNH3C

CAS RN 315703-52-7

IUPAC Name: N-(4-ethoxyphenyl)-4-{2-methylimidazo[1,2-a]pyridin-3-yl}-1,3-thiazol-2-amine
Synonyms: N-(4-ethoxyphenyl)-4-{2-methylimidazo[1,2-a]pyridin-3-yl}-1,3-thiazol-2-amine
Molecular Weight (g/mol): 350.44
Molecular Formula: C19H18N4OS
InChi Key: ROYXIPOUVGDTAO-UHFFFAOYSA-N
SMILES: CCOC1=CC=C(NC2=NC(=CS2)C2=C(C)N=C3C=CC=CN23)C=C1
Grade 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (4)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)