accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COOOCH3Br

CAS RN 31558-41-5

IUPAC Name: 4-bromo-2,5-dimethoxybenzaldehyde
Synonyms: 4-bromo-2,5-dimethoxybenzaldehyde
Molecular Weight (g/mol): 245.07
Molecular Formula: C9H9BrO3
InChi Key: BIFWGDWGCZLCHF-UHFFFAOYSA-N
SMILES: COC1=CC(Br)=C(OC)C=C1C=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results