accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COOCl

CAS RN 3153-36-4

IUPAC Name: ethyl 4-chlorobutanoate
Synonyms: ethyl 4-chlorobutanoate
Molecular Weight (g/mol): 150.6
Molecular Formula: C6H11ClO2
InChi Key: OPXNFHAILOHHFO-UHFFFAOYSA-N
SMILES: CCOC(=O)CCCCl
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (1)
  • (3)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (5)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)