accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

HClSnH

CAS RN 3151-41-5

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 429.58
Molecular Formula: C21H23ClSn
InChi Key: LHOUMPNYDACNQY-UHFFFAOYSA-N
SMILES: Cl.C([SnH](CC1=CC=CC=C1)CC1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)