accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 315-31-1

CH3FOHO

CAS RN 315-31-1

IUPAC Name: 2-fluoro-3-methylbenzoic acid
Synonyms: 2-fluoro-3-methylbenzoic acid
Molecular Weight (g/mol): 154.14
Molecular Formula: C8H7FO2
InChi Key: DGNAETGARNTCIL-UHFFFAOYSA-N
SMILES: CC1=CC=CC(C(O)=O)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results