accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

FFFNOSS

CAS RN 315-08-2

IUPAC Name: 2-sulfanylidene-3-[3-(trifluoromethyl)phenyl]-1,3-thiazolidin-4-one
Synonyms: 2-sulfanylidene-3-[3-(trifluoromethyl)phenyl]-1,3-thiazolidin-4-one
Molecular Weight (g/mol): 277.28
Molecular Formula: C10H6F3NOS2
InChi Key: MRDJXDJVVCNDND-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=CC(=C1)N1C(=O)CSC1=S
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results