accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OHOONHClCl(E)

CAS RN 31460-32-9

IUPAC Name: (2E)-3-[(2,5-dichlorophenyl)carbamoyl]prop-2-enoic acid
Synonyms: (2E)-3-[(2,5-dichlorophenyl)carbamoyl]prop-2-enoic acid
Molecular Weight (g/mol): 260.07
Molecular Formula: C10H7Cl2NO3
InChi Key: WSKTYRGKGHEEDM-ONEGZZNKSA-N
SMILES: OC(=O)\C=C\C(=O)NC1=CC(Cl)=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results