accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 31437-20-4

IUPAC Name: (pyrimidin-2-yl)thiourea
Synonyms: pyrimidin-2-ylthiourea
Molecular Weight (g/mol): 154.19
Molecular Formula: C5H6N4S
InChi Key: MCCLFFQZNBEOAV-UHFFFAOYSA-N
SMILES: NC(=S)NC1=NC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results