accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 314245-33-5

CH3NFH3CON

CAS RN 314245-33-5

IUPAC Name: 1-[1-(4-fluorophenyl)-2,5-dimethyl-1H-pyrrol-3-yl]-2-(pyrrolidin-1-yl)ethan-1-one
Synonyms: 1-[1-(4-fluorophenyl)-2,5-dimethylpyrrol-3-yl]-2-(pyrrolidin-1-yl)ethanone
Molecular Weight (g/mol): 300.38
Molecular Formula: C18H21FN2O
InChi Key: JUWDSDKJBMFLHE-UHFFFAOYSA-N
SMILES: CC1=CC(C(=O)CN2CCCC2)=C(C)N1C1=CC=C(F)C=C1
Grade 
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results