CAS RN 314054-21-2
CAS RN 314054-21-2
IUPAC Name:
2,6-difluoro-N-(2-phenylethyl)benzamide
Synonyms:
2,6-difluoro-N-(2-phenylethyl)benzamide
Molecular Weight (g/mol):
261.27
Molecular Formula:
C15H13F2NO
InChi Key:
PORBEPWKQBWEGX-UHFFFAOYSA-N
SMILES:
FC1=CC=CC(F)=C1C(=O)NCCC1=CC=CC=C1
1
–
1
of
1
results
1
2,6-Difluoro-N-(2-phenylethyl)benzamide, 97%, Thermo Scientific™
CAS: 314054-21-2 Molecular Formula: C15H13F2NO Molecular Weight (g/mol): 261.272 MDL Number: MFCD01175919 InChI Key: PORBEPWKQBWEGX-UHFFFAOYSA-N Synonym: 2,6-difluoro-n-2-phenylethyl benzamide,2,6-difluoro-n-phenethylbenzamide,2,6-difluoro-n-phenethyl-benzamide,2,6-difluoro-n-phenethylbenzenecarboxamide,2,6-difluorophenyl-n-2-phenylethyl carboxamide PubChem CID: 772213 IUPAC Name: 2,6-difluoro-N-(2-phenylethyl)benzamide SMILES: C1=CC=C(C=C1)CCNC(=O)C2=C(C=CC=C2F)F