accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 314-19-2

ClCH3N(R)OHOHH

CAS RN 314-19-2

IUPAC Name: hydrogen (9R)-10-methyl-10-azatetracyclo[7.7.1.0²,⁷.0¹³,¹⁷]heptadeca-1(17),2,4,6,13,15-hexaene-3,4-diol chloride
Synonyms: apomorphine hydrochloride
Molecular Weight (g/mol): 303.79
Molecular Formula: C17H18ClNO2
InChi Key: SKYZYDSNJIOXRL-BTQNPOSSSA-N
SMILES: [H+].[Cl-].CN1CCC2=CC=CC3=C2[C@H]1CC1=CC=C(O)C(O)=C31

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results