accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 31364-42-8

ONOONH2CCH2OO

CAS RN 31364-42-8

IUPAC Name: 4,7,13,16,21-pentaoxa-1,10-diazabicyclo[8.8.5]tricosane
Synonyms: 4,7,13,16,21-pentaoxa-1,10-diazabicyclo[8.8.5]tricosane
Molecular Weight (g/mol): 332.44
Molecular Formula: C16H32N2O5
InChi Key: HDLXPNDSLDLJHF-UHFFFAOYSA-N
SMILES: C1COCCN2CCOCCOCCN(CCO1)CCOCC2
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results