accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 313546-16-6

BOH3CFFFHOOH

CAS RN 313546-16-6

IUPAC Name: [4-methoxy-2-(trifluoromethyl)phenyl]boronic acid
Synonyms: 4-methoxy-2-(trifluoromethyl)phenylboronic acid
Molecular Weight (g/mol): 219.95
Molecular Formula: C8H8BF3O3
InChi Key: ZBCRZEJNAADYKG-UHFFFAOYSA-N
SMILES: COC1=CC(=C(C=C1)B(O)O)C(F)(F)F
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results