accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 313545-34-5

BFFFFHOOH

CAS RN 313545-34-5

IUPAC Name: [2-fluoro-6-(trifluoromethyl)phenyl]boronic acid
Synonyms: 2-fluoro-6-(trifluoromethyl)phenylboronic acid
Molecular Weight (g/mol): 207.92
Molecular Formula: C7H5BF4O2
InChi Key: TXBRJTKFAYXKMV-UHFFFAOYSA-N
SMILES: OB(O)C1=C(C=CC=C1F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results