accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

BClCH3HOOH

CAS RN 313545-20-9

IUPAC Name: (3-chloro-2-methylphenyl)boronic acid
Synonyms: 3-chloro-2-methylphenylboronic acid
Molecular Weight (g/mol): 170.4
Molecular Formula: C7H8BClO2
InChi Key: BJPNVVXTUYMJPN-UHFFFAOYSA-N
SMILES: CC1=C(C=CC=C1Cl)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results