CAS RN 313493-80-0
CAS RN 313493-80-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
2
CID 1375606, MedChemExpress
MedChemExpress CID 1375606 is a surrogate agonist of orphan G protein-coupled receptor GPR27.