accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 3133-01-5

H3COH

CAS RN 3133-01-5

IUPAC Name: tricosan-1-ol
Synonyms: tricosan-1-ol
Molecular Weight (g/mol): 340.64
Molecular Formula: C23H48O
InChi Key: FPLNRAYTBIFSFW-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCCCCO
Boiling Point 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)