accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 313241-14-4

NNOOHFFF

CAS RN 313241-14-4

IUPAC Name: 2-[2-(trifluoromethyl)-1H-1,3-benzodiazol-1-yl]acetic acid
Synonyms: [2-(trifluoromethyl)-1,3-benzodiazol-1-yl]acetic acid
Molecular Weight (g/mol): 244.17
Molecular Formula: C10H7F3N2O2
InChi Key: GPOYZFXQCCJBTF-UHFFFAOYSA-N
SMILES: OC(=O)CN1C2=CC=CC=C2N=C1C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results