CAS RN 31314-39-3
CAS RN 31314-39-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Prodipine hydrochloride, MedChemExpress
MedChemExpress Prodipine, a diphenyl-phosphonate derivative. The IC50s of Prodipine for purified and plasma Dipeptidyl peptidase IV (DPP IV) from the rabbit are 4.5 μM and 30 μM, respectively.