accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 312946-37-5

SONNSOHNNH2

CAS RN 312946-37-5

IUPAC Name: 2-({4-oxo-3,5-diphenyl-3H,4H-thieno[2,3-d]pyrimidin-2-yl}sulfanyl)acetohydrazide
Synonyms: 2-({4-oxo-3,5-diphenylthieno[2,3-d]pyrimidin-2-yl}sulfanyl)acetohydrazide
Molecular Weight (g/mol): 408.49
Molecular Formula: C20H16N4O2S2
InChi Key: WLBUICQBNZXIDJ-UHFFFAOYSA-N
SMILES: NNC(=O)CSC1=NC2=C(C(=CS2)C2=CC=CC=C2)C(=O)N1C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Color 
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results