accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 31284-89-6

CH3OHOOH

CAS RN 31284-89-6

IUPAC Name: 2-hydroxy-2-(4-methylphenyl)acetic acid
Synonyms: hydroxy(4-methylphenyl)acetic acid
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: SFGURAWGCAPHON-UHFFFAOYNA-N
SMILES: CC1=CC=C(C=C1)C(O)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results