accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 3128-07-2

H3COOHO

CAS RN 3128-07-2

IUPAC Name: 6-oxoheptanoic acid
Synonyms: 5-aceto valeric acid
Molecular Weight (g/mol): 144.17
Molecular Formula: C7H12O3
InChi Key: IZOQMUVIDMLRDC-UHFFFAOYSA-N
SMILES: CC(=O)CCCCC(O)=O
Boiling Point 
  • (6)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (6)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)