accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 31277-98-2

PdPPPPPhPhPhPhPhPhPhPh

CAS RN 31277-98-2

IUPAC Name: bis([2-(diphenylphosphanyl)ethyl]diphenylphosphane) palladium
Synonyms: bis(diphos) palladium
Molecular Weight (g/mol): 903.27
Molecular Formula: C52H48P4Pd
InChi Key: FAFGMAGIYHHRKN-UHFFFAOYSA-N
SMILES: [Pd].C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results