accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 312745-91-8

H3COONOH

CAS RN 312745-91-8

IUPAC Name: ethyl 4-cyano-3-hydroxybutanoate
Synonyms: ethyl 4-cyano-3-hydroxybutanoate
Molecular Weight (g/mol): 157.17
Molecular Formula: C7H11NO3
InChi Key: LOQFROBMBSKWQY-UHFFFAOYNA-N
SMILES: CCOC(=O)CC(O)CC#N
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results