accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 31272-04-5

CH3NNOCH3

CAS RN 31272-04-5

IUPAC Name: 1-methyl-3-propyl-4,5-dihydro-1H-pyrazol-5-one
Synonyms: 2-methyl-5-propyl-4H-pyrazol-3-one
Molecular Weight (g/mol): 140.19
Molecular Formula: C7H12N2O
InChi Key: RULINAPACRUGOF-UHFFFAOYSA-N
SMILES: CCCC1=NN(C)C(=O)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results