accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 31270-80-1

NOCl

CAS RN 31270-80-1

IUPAC Name: 4-chlorofuro[3,2-c]pyridine
Synonyms: 4-chlorofuro[3,2-c]pyridine
Molecular Weight (g/mol): 153.57
Molecular Formula: C7H4ClNO
InChi Key: OPFFYLJLHPZSEO-UHFFFAOYSA-N
SMILES: ClC1=C2C=COC2=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results