accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 312693-72-4

NN(R)NHNO(R)(S)OH2NHOHOH2O

CAS RN 312693-72-4

IUPAC Name: 2-amino-9-[4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-3H-purin-6-one hydrate
Synonyms: deoxyguanosine hydrate
Molecular Weight (g/mol): 285.26
Molecular Formula: C10H15N5O5
InChi Key: LZSCQUCOIRGCEJ-UHFFFAOYNA-N
SMILES: O.NC1=NC(=O)C2=C(N1)N(C=N2)C1CC(O)C(CO)O1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)