accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CZnBr

CAS RN 312624-15-0

IUPAC Name: adamantan-1-ide; bromozincylium
Synonyms: adamantan-1-ide; bromozincylium
Molecular Weight (g/mol): 280.51
Molecular Formula: C10H15BrZn
InChi Key: PTWVHBHYYPLVCD-UHFFFAOYSA-M
SMILES: [Zn+]Br.C1C2CC3CC1C[C-](C2)C3

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results