accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 312300-42-8

H3CONSOOCl

CAS RN 312300-42-8

IUPAC Name: 6-methoxypyridine-3-sulfonyl chloride
Synonyms: 6-methoxypyridine-3-sulfonyl chloride
Molecular Weight (g/mol): 207.63
Molecular Formula: C6H6ClNO3S
InChi Key: OMLIVJOTRYWHNY-UHFFFAOYSA-N
SMILES: COC1=NC=C(C=C1)S(Cl)(=O)=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results