accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 31224-82-5

NFFFO

CAS RN 31224-82-5

IUPAC Name: 5-(trifluoromethyl)pyridine-2-carbaldehyde
Synonyms: 5-(trifluoromethyl)pyridine-2-carbaldehyde
Molecular Weight (g/mol): 175.11
Molecular Formula: C7H4F3NO
InChi Key: JJDDVGAESNBKMY-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CN=C(C=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results