accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 31181-90-5

BrON

CAS RN 31181-90-5

IUPAC Name: 5-bromopyridine-2-carbaldehyde
Synonyms: 5-bromopyridine-2-carbaldehyde
Molecular Weight (g/mol): 186.01
Molecular Formula: C6H4BrNO
InChi Key: ZQVLPMNLLKGGIU-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=O)N=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)