accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 3115-66-0

IH3CPCH3H3CH3C

CAS RN 3115-66-0

IUPAC Name: tetrabutylphosphanium iodide
Synonyms: tetrabutylphosphonium iodide
Molecular Weight (g/mol): 386.34
Molecular Formula: C16H36IP
InChi Key: CCIYPTIBRAUPLQ-UHFFFAOYSA-M
SMILES: [I-].CCCC[P+](CCCC)(CCCC)CCCC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results