accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 31139-36-3

OOOOO

CAS RN 31139-36-3

IUPAC Name: dibenzyl dicarbonate
Synonyms: dibenzyl dicarbonate
Molecular Weight (g/mol): 286.28
Molecular Formula: C16H14O5
InChi Key: FHRRJZZGSJXPRQ-UHFFFAOYSA-N
SMILES: O=C(OCC1=CC=CC=C1)OC(=O)OCC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results