accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 31080-39-4

H3CCH3OHO

CAS RN 31080-39-4

IUPAC Name: 2-propylheptanoic acid
Synonyms: 2-propylheptanoic acid
Molecular Weight (g/mol): 172.27
Molecular Formula: C10H20O2
InChi Key: RXGPYPPCEXISOV-UHFFFAOYNA-N
SMILES: CCCCCC(CCC)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results