accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 30992-29-1

H3CH3CCH3OONHH3CCH3OO

CAS RN 30992-29-1

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-2-methylpropanoate
Synonyms: 2-[(tert-butoxycarbonyl)amino]-2-methylpropanoate
Molecular Weight (g/mol): 202.23
Molecular Formula: C9H16NO4
InChi Key: MFNXWZGIFWJHMI-UHFFFAOYSA-M
SMILES: CC(C)(C)OC(=O)NC(C)(C)C([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)