accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 309913-83-5

CH3NNONH3COONH3CCH3ClCH3F

CAS RN 309913-83-5

IUPAC Name: 2-(6-chloro-5-{4-[(4-fluorophenyl)methyl]-2,5-dimethylpiperazine-1-carbonyl}-1-methyl-1H-indol-3-yl)-N,N-dimethyl-2-oxoacetamide
Synonyms: 2-(6-chloro-5-{4-[(4-fluorophenyl)methyl]-2,5-dimethylpiperazine-1-carbonyl}-1-methylindol-3-yl)-N,N-dimethyl-2-oxoacetamide
Molecular Weight (g/mol): 513.01
Molecular Formula: C27H30ClFN4O3
InChi Key: ZMELOYOKMZBMRB-UHFFFAOYNA-N
SMILES: CC1CN(C(C)CN1CC1=CC=C(F)C=C1)C(=O)C1=C(Cl)C=C2N(C)C=C(C(=O)C(=O)N(C)C)C2=C1
Grade 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results