accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 30984-28-2

CH3CH3OHOFFF(E)

CAS RN 30984-28-2

IUPAC Name: (4E)-6,6,6-trifluoro-5-hydroxy-2-methylhex-4-en-3-one
Synonyms: (4E)-6,6,6-trifluoro-5-hydroxy-2-methylhex-4-en-3-one
Molecular Weight (g/mol): 182.14
Molecular Formula: C7H9F3O2
InChi Key: QDJWKASBKAMIDF-ZZXKWVIFSA-N
SMILES: CC(C)C(=O)\C=C(\O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results