CAS RN 3096-47-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Chloro-9-fluorenone, TRC
CAS: 3096-47-7 Molecular Formula: C13 H7 Cl O Molecular Weight (g/mol): 214.65 Synonym: 2-Chloro-9H-fluoren-9-one,2-Chloro-9-fluorenone,2-Chlorofluorenone,NSC 107566 IUPAC Name: 2-chlorofluoren-9-one SMILES: Clc1ccc2c3ccccc3C(=O)c2c1