accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

AlH3COCH3OCH3O

CAS RN 3085-30-1

IUPAC Name: aluminium(3+) tris(butan-1-olate)
Synonyms: aluminium(3+) tris(butan-1-olate)
Molecular Weight (g/mol): 246.33
Molecular Formula: C12H27AlO3
InChi Key: MYWQGROTKMBNKN-UHFFFAOYSA-N
SMILES: [Al+3].CCCC[O-].CCCC[O-].CCCC[O-]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results