accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 3080-99-7

IUPAC Name: 3,4-dihydro-2H-1,4-benzothiazine
Synonyms: 3,4-dihydro-2H-1,4-benzothiazine
Molecular Weight (g/mol): 151.23
Molecular Formula: C8H9NS
InChi Key: YBBLSBDJIKMXNQ-UHFFFAOYSA-N
SMILES: C1CSC2=CC=CC=C2N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results