CAS RN 307310-74-3
CAS RN 307310-74-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Lithium periodate dihydrate, Reagent Grade
CAS: 307310-74-3 Molecular Formula: ILiO4 Molecular Weight (g/mol): 197.84 MDL Number: MFCD00049474 InChI Key: SYWXNZXEJFSLEU-UHFFFAOYSA-M Synonym: lithium periodate,lithotab io4,acmc-20akme,lithium 1+ ion periodate,lithium 1+ periodate,lithium periodate lih4io6 PubChem CID: 23711816 IUPAC Name: lithium;periodate SMILES: [Li+].[O-]I(=O)(=O)=O