accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NNFFFOHO

CAS RN 306960-77-0

IUPAC Name: 2-(trifluoromethyl)pyrimidine-5-carboxylic acid
Synonyms: 2-(trifluoromethyl)pyrimidine-5-carboxylic acid
Molecular Weight (g/mol): 192.1
Molecular Formula: C6H3F3N2O2
InChi Key: RGLZXVNOHDUMRJ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CN=C(N=C1)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results