accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 306936-90-3

CH3H3CCH3NNHSO

CAS RN 306936-90-3

IUPAC Name: 5-(4-tert-butylphenyl)-2,3-dihydro-1,3,4-oxadiazole-2-thione
Synonyms: 5-(4-tert-butylphenyl)-3H-1,3,4-oxadiazole-2-thione
Molecular Weight (g/mol): 234.32
Molecular Formula: C12H14N2OS
InChi Key: ATIYHEZPBLMRNI-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(C=C1)C1=NNC(=S)O1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results