accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 306936-20-9

IUPAC Name: 2-methyl-1-[3-(morpholin-4-yl)propyl]-5-phenyl-1H-pyrrole-3-carboxylic acid
Synonyms: 2-methyl-1-[3-(morpholin-4-yl)propyl]-5-phenylpyrrole-3-carboxylic acid
Molecular Weight (g/mol): 328.41
Molecular Formula: C19H24N2O3
InChi Key: OZRPKALPSGYEHX-UHFFFAOYSA-N
SMILES: CC1=C(C=C(N1CCCN1CCOCC1)C1=CC=CC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results