accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

FFFClOO

CAS RN 306936-04-9

IUPAC Name: 5-[2-chloro-4-(trifluoromethyl)phenyl]furan-2-carbaldehyde
Synonyms: 5-[2-chloro-4-(trifluoromethyl)phenyl]furan-2-carbaldehyde
Molecular Weight (g/mol): 274.62
Molecular Formula: C12H6ClF3O2
InChi Key: SAEPEPVLSICNRE-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(Cl)=C(C=C1)C1=CC=C(O1)C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results