accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OFFFSOOCl

CAS RN 306935-02-4

IUPAC Name: 5-methyl-2-(trifluoromethyl)furan-3-sulfonyl chloride
Synonyms: 5-methyl-2-(trifluoromethyl)furan-3-sulfonyl chloride
Molecular Weight (g/mol): 248.6
Molecular Formula: C6H4ClF3O3S
InChi Key: GSYXWDYWBRICSD-UHFFFAOYSA-N
SMILES: CC1=CC(=C(O1)C(F)(F)F)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results